(Cyclohexylmethyl)hydrazine hydrochloride 97%

Proveedor: Apollo Scientific
Atención

Sinónimos: (Hydrazinomethyl)cyclohexane hydrochloride

OR0199-1G OR0199-5G OR0199-10G
APOSOR0199-1GEA 102 EUR
APOSOR0199-1G APOSOR0199-5G APOSOR0199-10G
(Cyclohexylmethyl)hydrazine hydrochloride 97%
(Cyclohexylmethyl)hydrazine hydrochloride
Formula: C₇H₁₆N₂·ClH
MW: 164,676 g/mol
Pto. de fusión: 141…144 °C
Temperatura de almacenaje: Nevera
Número MDL: MFCD06245509
Núm. CAS: 3637-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR