1-{2-[(2-Chlorobenzyl)thio]ethyl}-5-oxopyrrolidine-3-carboxylic acid
Proveedor: Apollo Scientific
OR110083-1G
OR110083-10G
OR110083-5G
APOSOR110083-1GEA
310
EUR
APOSOR110083-1G
APOSOR110083-10G
APOSOR110083-5G
1-{2-[(2-Chlorobenzyl)thio]ethyl}-5-oxopyrrolidine-3-carboxylic acid
1-{2-[(2-Chlorobenzyl)thio]ethyl}-5-oxopyrrolidine-3-carboxylic acid
Formula:
C₁₄H₁₆ClNO₃S MW: 313,8 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD23381091 Núm. CAS: 1440535-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...