2,2,6,6-Tetramethyl-1-(4-nitrobenzoyl)piperidine
Proveedor: Apollo Scientific
OR111115-1G
OR111115-5G
APOSOR111115-1GEA
0
EUR
APOSOR111115-1G
APOSOR111115-5G
2,2,6,6-Tetramethyl-1-(4-nitrobenzoyl)piperidine
2,2,6,6-Tetramethyl-1-(4-nitrobenzoyl)piperidine
Formula:
C₁₆H₂₂N₂O₃ MW: 290,357 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28954642 Núm. CAS: 213414-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...