3-(2-Aminoethyl)azetidine hydrochloride, 3-BOC protected

Proveedor: Apollo Scientific
Atención

Sinónimos: tert-butyl N-[2-(azetidin-3-yl)ethyl]carbamate hydrochloride, tert-Butyl [2-(azetidin-3-yl)ethyl]carbamate hydrochloride, tert-Butyl (2-(azetidin-3-yl)ethyl)carbamate hydrochloride

OR2246-100MG OR2246-1G
APOSOR2246-100MGEA 69.6 EUR
APOSOR2246-100MG APOSOR2246-1G
3-(2-Aminoethyl)azetidine hydrochloride, 3-BOC protected
3-(2-Aminoethyl)azetidine hydrochloride, 3-BOC protected
Formula: C₁₀H₂₀N₂O₂·ClH
MW: 236,739 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09998720
Núm. CAS: 1170905-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR