3-Amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylic acid
Proveedor: Apollo Scientific
OR300746-1G
OR300746-250MG
OR300746-500MG
OR300746-2.5G
OR300746-5G
APOSOR300746-1GEA
249
EUR
APOSOR300746-1G
APOSOR300746-250MG
APOSOR300746-500MG
APOSOR300746-2.5G
APOSOR300746-5G
3-Amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylic acid
3-Amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylic acid
Formula:
C₁₀H₁₀N₂O₂S Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01085687 Núm. CAS: 58327-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...