Ethyl 4,5-dimethyl-1,3-thiazole-2-carboxylate
Proveedor: Apollo Scientific
OR303129-1G
OR303129-500MG
APOSOR303129-1GEA
270
EUR
APOSOR303129-1G
APOSOR303129-500MG
Ethyl 4,5-dimethyl-1,3-thiazole-2-carboxylate
Ethyl 4,5-dimethyl-1,3-thiazole-2-carboxylate
Formula:
C₈H₁₁NO₂S MW: 185,243 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD14654437 Núm. CAS: 79247-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...