(8S)-8-Amino-6-azaspiro[3.4]octan-5-one
Proveedor: Apollo Scientific
OR304435-100MG
OR304435-250MG
OR304435-500MG
APOSOR304435-100MGEA
388
EUR
APOSOR304435-100MG
APOSOR304435-250MG
APOSOR304435-500MG
(8S)-8-Amino-6-azaspiro[3.4]octan-5-one
(8S)-8-Amino-6-azaspiro[3.4]octan-5-one
Formula:
C₇H₁₂N₂O MW: 140,183 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD29037455 Núm. CAS: 1810074-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...