diexo-3-Amino-bicyclo[2.2.1]heptane-2-carboxylic acid amide
Proveedor: Apollo Scientific
OR307550-1G
OR307550-250MG
APOSOR307550-1GEA
912
EUR
APOSOR307550-1G
APOSOR307550-250MG
diexo-3-Amino-bicyclo[2.2.1]heptane-2-carboxylic acid amide
diexo-3-Amino-bicyclo[2.2.1]heptane-2-carboxylic acid amide
Formula:
C₈H₁₄N₂O MW: 154,21 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07783735 Núm. CAS: 105786-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...