6-Bromo-4-hydrazinoquinoline hydrochloride
Proveedor: Apollo Scientific
OR307824-1G
OR307824-5G
APOSOR307824-1GEA
301
EUR
APOSOR307824-1G
APOSOR307824-5G
6-Bromo-4-hydrazinoquinoline hydrochloride
6-Bromo-4-hydrazinoquinoline hydrochloride
Formula:
C₉H₈BrN₃·ClH MW: 274,545 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08437007 Núm. CAS: 1171943-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...