2,7,8-Trimethylquinoline-3-carboxylic acid
Proveedor: Apollo Scientific
OR307854-1G
OR307854-5G
APOSOR307854-1GEA
325
EUR
APOSOR307854-1G
APOSOR307854-5G
2,7,8-Trimethylquinoline-3-carboxylic acid
2,7,8-Trimethylquinoline-3-carboxylic acid
Formula:
C₁₃H₁₃NO₂ MW: 215,248 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08437591 Núm. CAS: 92513-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...