7-Methylquinoline-3-carboxylic acid
Proveedor: Apollo Scientific
OR309231-1G
OR309231-5G
APOSOR309231-1GEA
393
EUR
APOSOR309231-1G
APOSOR309231-5G
7-Methylquinoline-3-carboxylic acid
7-Methylquinoline-3-carboxylic acid
Formula:
C₁₁H₉NO₂ MW: 187,195 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09787877 Núm. CAS: 948291-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...