Methyl 3-formyl-4-methoxybenzoate
Proveedor: Apollo Scientific
OR310929-1G
OR310929-500MG
APOSOR310929-1GEA
538
EUR
APOSOR310929-1G
APOSOR310929-500MG
Methyl 3-formyl-4-methoxybenzoate
Methyl 3-formyl-4-methoxybenzoate
Formula:
C₁₀H₁₀O₄ MW: 194,184 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09832059 Núm. CAS: 145742-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...