2-(3-Ethenylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Proveedor: Apollo Scientific
OR310976-1G
OR310976-5G
APOSOR310976-1GEA
53.8
EUR
APOSOR310976-1G
APOSOR310976-5G
2-(3-Ethenylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(3-Ethenylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula:
C₁₄H₁₉BO₂ MW: 230,11 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD18733869 Núm. CAS: 627525-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...