3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid
Proveedor: Apollo Scientific
OR312451-250MG
APOSOR312451-250MGEA
502
EUR
APOSOR312451-250MG
3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid
3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid
Formula:
C₇H₁₀O₂ MW: 126,153 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD19228632 Núm. CAS: 65862-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...