(1-Benzyl-1H-imidazol-5-yl)methanamine
Proveedor: Apollo Scientific
OR321524-1G
OR321524-250MG
APOSOR321524-1GEA
719
EUR
APOSOR321524-1G
APOSOR321524-250MG
(1-Benzyl-1H-imidazol-5-yl)methanamine
(1-Benzyl-1H-imidazol-5-yl)methanamine
Formula:
C₁₁H₁₃N₃ MW: 187,241 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD21703522 Núm. CAS: 1439991-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...