4-Hydroxyquinoline-6-carbonitrile 96%
Proveedor: Apollo Scientific
OR350490-1G
OR350490-250MG
APOSOR350490-1GEA
232
EUR
APOSOR350490-1G
APOSOR350490-250MG
4-Hydroxyquinoline-6-carbonitrile 96%
4-Hydroxyquinoline-6-carbonitrile
Formula:
C₁₀H₆N₂O MW: 170,167 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD08063203 Núm. CAS: 219763-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...