3-Acetamidophenacyl bromide
Proveedor: Apollo Scientific
|
Peligro
|
OR350581-1G
OR350581-250MG
OR350581-5G
APOSOR350581-1GEA
0
EUR
APOSOR350581-1G
APOSOR350581-250MG
APOSOR350581-5G
3-Acetamidophenacyl bromide
3-Acetamidophenacyl bromide
Formula:
C₁₀H₁₀BrNO₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07364400 Núm. CAS: 30095-56-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...