7-Bromo-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole ≥98%
Proveedor: Apollo Scientific
OR361740-100MG
OR361740-1G
OR361740-250MG
APOSOR361740-100MGEA
319
EUR
APOSOR361740-100MG
APOSOR361740-1G
APOSOR361740-250MG
7-Bromo-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole ≥98%
7-Bromo-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
Formula:
C₁₁H₁₁BrN₂ MW: 251,122 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD21337853 Núm. CAS: 108061-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...