2-Methyl-octahydro-pyrrolo[3,4-c]pyrrole ≥98%
Proveedor: Apollo Scientific
OR370041-100MG
OR370041-250MG
APOSOR370041-100MGEA
52.2
EUR
APOSOR370041-100MG
APOSOR370041-250MG
2-Methyl-octahydro-pyrrolo[3,4-c]pyrrole ≥98%
2-Methyl-octahydro-pyrrolo[3,4-c]pyrrole
Formula:
C₇H₁₄N₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09608113 Núm. CAS: 86732-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...