Methyl 4-bromo-3-(chlorosulphonyl)benzoate
Proveedor: Apollo Scientific
|
Peligro
|
OR42049-1G
OR42049-250MG
OR42049-5G
APOSOR42049-1GEA
223
EUR
APOSOR42049-1G
APOSOR42049-250MG
APOSOR42049-5G
Methyl 4-bromo-3-(chlorosulphonyl)benzoate
Methyl 4-bromo-3-(chlorosulphonyl)benzoate
Formula:
C₈H₆BrClO₄S MW: 313,553 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD12169456 Núm. CAS: 168969-13-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...