4-Iminothiazol-2-amine
Proveedor: Apollo Scientific
OR450097-1G
OR450097-250MG
APOSOR450097-1GEA
429
EUR
APOSOR450097-1G
APOSOR450097-250MG
4-Iminothiazol-2-amine
4-Iminothiazol-2-amine
Formula:
C₃H₅N₃S MW: 115,157 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01680852 Núm. CAS: 26246-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...