8-Bromoindolo[2,1-b]quinazoline-6,12-dione
Proveedor: Apollo Scientific
OR450114-1G
OR450114-250MG
APOSOR450114-1GEA
429
EUR
APOSOR450114-1G
APOSOR450114-250MG
8-Bromoindolo[2,1-b]quinazoline-6,12-dione
8-Bromoindolo[2,1-b]quinazoline-6,12-dione
Formula:
C₁₅H₇BrN₂O₂ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07637587 Núm. CAS: 65618-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...