3-Chlorobenzoyl isocyanate

Proveedor: Apollo Scientific

OR450132-1G OR450132-250MG
APOSOR450132-1GEA 509 EUR
APOSOR450132-1G APOSOR450132-250MG
3-Chlorobenzoyl isocyanate
3-Chlorobenzoyl isocyanate

May form small amount of sediment

Formula: C₈H₄ClNO₂
MW: 181,576 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD09817215
Núm. CAS: 74703-17-6
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR