1-Cyclobutyl-4-piperidinamine dihydrochloride 98%
Proveedor: Apollo Scientific
OR451075-5G
OR451075-1G
APOSOR451075-5GEA
3470
EUR
APOSOR451075-5G
APOSOR451075-1G
1-Cyclobutyl-4-piperidinamine dihydrochloride 98%
1-Cyclobutyl-4-piperidinamine dihydrochloride
Formula:
C₉H₁₈N₂·2ClH MW: 227,175 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09759200 Núm. CAS: 1048921-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...