Fmoc-DL-phenylglycine 97%

Proveedor: Apollo Scientific

OR452093-1G OR452093-25G OR452093-5G
APOSOR452093-1GEA 98.6 EUR
APOSOR452093-1G APOSOR452093-25G APOSOR452093-5G
Fmoc-DL-phenylglycine 97%
Fmoc-DL-phenylglycine
Formula: C₂₃H₁₉NO₄
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02094475
Núm. CAS: 163883-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR