(3-Bromophenyl)-4-piperidinyl-methanone hydrochloride 97%
Proveedor: Apollo Scientific
OR452129-1G
OR452129-250MG
OR452129-500MG
APOSOR452129-1GEA
806
EUR
APOSOR452129-1G
APOSOR452129-250MG
APOSOR452129-500MG
(3-Bromophenyl)-4-piperidinyl-methanone hydrochloride 97%
(3-Bromophenyl)-4-piperidinyl-methanone hydrochloride
Formula:
C₁₂H₁₄BrNO·ClH MW: 304,611 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11846278 Núm. CAS: 1225573-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...