3-(3-Pyridyl)-D-alanine dihydrochloride
Proveedor: Apollo Scientific
OR460015-1G
OR460015-250MG
APOSOR460015-1GEA
372
EUR
APOSOR460015-1G
APOSOR460015-250MG
3-(3-Pyridyl)-D-alanine dihydrochloride
3-(3-Pyridyl)-D-alanine dihydrochloride
Formula:
C₈H₁₀N₂O₂·2ClH Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00191179 Núm. CAS: 93960-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...