2-(4-(Dimethylamino)phenyl)acetyl chloride
Proveedor: Apollo Scientific
OR46679-1G
OR46679-250MG
OR46679-500MG
APOSOR46679-1GEA
1040
EUR
APOSOR46679-1G
APOSOR46679-250MG
APOSOR46679-500MG
2-(4-(Dimethylamino)phenyl)acetyl chloride
2-(4-(Dimethylamino)phenyl)acetyl chloride
Formula:
C₁₀H₁₂ClNO Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD27950360 Núm. CAS: 39889-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...