Methyl 2-Bromo-5-sulphamoylbenzoate

Proveedor: Apollo Scientific

OR470123-1G OR470123-5G
APOSOR470123-1GEA 397 EUR
APOSOR470123-1G APOSOR470123-5G
Methyl 2-Bromo-5-sulphamoylbenzoate
Methyl 2-Bromo-5-sulphamoylbenzoate
Formula: C₈H₈BrNO₄S
MW: 294,122 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11057846
Núm. CAS: 924867-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR