(1S,6S)-7-Cbz-3,7-diazabicyclo[4.2.0]octane
Proveedor: Apollo Scientific
OR470484-1G
OR470484-250MG
APOSOR470484-1GEA
2860
EUR
APOSOR470484-1G
APOSOR470484-250MG
(1S,6S)-7-Cbz-3,7-diazabicyclo[4.2.0]octane
(1S,6S)-7-Cbz-3,7-diazabicyclo[4.2.0]octane
Formula:
C₁₄H₁₈N₂O₂ MW: 246,305 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22205856 Núm. CAS: 1932306-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...