4-Chlorohexanoyl chloride
Proveedor: Apollo Scientific
OR470809-5G
OR470809-1G
APOSOR470809-5GEA
804
EUR
APOSOR470809-5G
APOSOR470809-1G
4-Chlorohexanoyl chloride
4-Chlorohexanoyl chloride
Formula:
C₆H₁₀Cl₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD20622929 Núm. CAS: 99585-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...