2-Chloro-4-(cyclopropylmethoxy)benzoic acid
Proveedor: Apollo Scientific
OR470993-1G
OR470993-5G
APOSOR470993-1GEA
397
EUR
APOSOR470993-1G
APOSOR470993-5G
2-Chloro-4-(cyclopropylmethoxy)benzoic acid
2-Chloro-4-(cyclopropylmethoxy)benzoic acid
Formula:
C₁₁H₁₁ClO₃ MW: 226,656 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD17256604 Núm. CAS: 1237084-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...