3-Methylpyrrole-2,4-dicarboxylic acid
Proveedor: Apollo Scientific
OR471303-1G
OR471303-250MG
APOSOR471303-1GEA
601
EUR
APOSOR471303-1G
APOSOR471303-250MG
3-Methylpyrrole-2,4-dicarboxylic acid
3-Methylpyrrole-2,4-dicarboxylic acid
Formula:
C₇H₇NO₄ MW: 169,135 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09991921 Núm. CAS: 3780-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...