(S)-2-Phenylpropanal
Proveedor: Apollo Scientific
OR471313-1G
OR471313-250MG
APOSOR471313-1GEA
855
EUR
APOSOR471313-1G
APOSOR471313-250MG
(S)-2-Phenylpropanal
(S)-2-Phenylpropanal
Formula:
C₉H₁₀O MW: 134,175 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD24673692 Núm. CAS: 33530-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...