5-(2-Methoxy-phenyl)-2H-pyrazole-3-carboxylic acid methyl ester 95%
Proveedor: Apollo Scientific
|
Atención
|
OR480228-1G
OR480228-250MG
OR480228-500MG
APOSOR480228-1GEA
350
EUR
APOSOR480228-1G
APOSOR480228-250MG
APOSOR480228-500MG
5-(2-Methoxy-phenyl)-2H-pyrazole-3-carboxylic acid methyl ester 95%
5-(2-Methoxy-phenyl)-2H-pyrazole-3-carboxylic acid methyl ester
Formula:
C₁₂H₁₂N₂O₃ MW: 232,235 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03761321 Núm. CAS: 898052-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...