3-(1-Methylpyrazol-3-yl)propanoic acid 95%
Proveedor: Apollo Scientific
OR480239-1G
OR480239-250MG
OR480239-500MG
APOSOR480239-1GEA
931
EUR
APOSOR480239-1G
APOSOR480239-250MG
APOSOR480239-500MG
3-(1-Methylpyrazol-3-yl)propanoic acid 95%
3-(1-Methylpyrazol-3-yl)propanoic acid
Formula:
C₇H₁₀N₂O₂ MW: 154.166 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06808504 Núm. CAS: 1006440-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...