2-Amino-2-(4-tert-butylphenyl)acetic acid
Proveedor: Apollo Scientific
OR480452-1G
OR480452-2G
OR480452-5G
APOSOR480452-1GEA
171
EUR
APOSOR480452-1G
APOSOR480452-2G
APOSOR480452-5G
2-Amino-2-(4-tert-butylphenyl)acetic acid
2-Amino-2-(4-tert-butylphenyl)acetic acid
Formula:
C₁₂H₁₇NO₂ MW: 207,269 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02662470 Núm. CAS: 299165-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...