2-Amino-2-(2,4-dichlorophenyl)acetic acid hydrochloride
Proveedor: Apollo Scientific
OR480456-2G
OR480456-5G
APOSOR480456-2GEA
260
EUR
APOSOR480456-2G
APOSOR480456-5G
2-Amino-2-(2,4-dichlorophenyl)acetic acid hydrochloride
2-Amino-2-(2,4-dichlorophenyl)acetic acid hydrochloride
Formula:
C₈H₇Cl₂NO₂·ClH MW: 256,514 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04966782 Núm. CAS: 1031790-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...