2-Amino-2-(4-methoxy-3-methyl-phenyl)acetic acid
Proveedor: Apollo Scientific
OR480471-1G
OR480471-500MG
APOSOR480471-1GEA
451
EUR
APOSOR480471-1G
APOSOR480471-500MG
2-Amino-2-(4-methoxy-3-methyl-phenyl)acetic acid
2-Amino-2-(4-methoxy-3-methyl-phenyl)acetic acid
Formula:
C₁₀H₁₃NO₃ MW: 195,215 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02662436 Núm. CAS: 1024613-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...