1',3'-Dihydro-spiro(imidazolidine-4,2'-(2H)indene)-2,5-dione 97%
Proveedor: Apollo Scientific
OR480755-1G
OR480755-5G
APOSOR480755-1GEA
0
EUR
APOSOR480755-1G
APOSOR480755-5G
1',3'-Dihydro-spiro(imidazolidine-4,2'-(2H)indene)-2,5-dione 97%
1',3'-Dihydro-spiro(imidazolidine-4,2'-(2H)indene)-2,5-dione
Formula:
C₁₁H₁₀N₂O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01683900 Núm. CAS: 27473-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...