Methyl 4-phenylbutanoate 98%

Proveedor: Apollo Scientific

OR480836-5G OR480836-25G
APOSOR480836-5GEA 159 EUR
APOSOR480836-5G APOSOR480836-25G
Methyl 4-phenylbutanoate 98%
Methyl 4-phenylbutanoate
Formula: C₁₁H₁₄O₂
MW: 178,228 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00043756
Núm. CAS: 2046-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR