5-Cyclopropyl-2-thiophenesulphonyl chloride
Proveedor: Apollo Scientific
|
Peligro
|
OR48097-1G
OR48097-250MG
APOSOR48097-1GEA
116
EUR
APOSOR48097-1G
APOSOR48097-250MG
5-Cyclopropyl-2-thiophenesulphonyl chloride
5-Cyclopropyl-2-thiophenesulphonyl chloride
Formula:
C₇H₇ClO₂S₂ MW: 222,712 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD31665813 Núm. CAS: 2141875-46-7 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...