7-Nitro-1H-benzo[d][1,2,3]triazole-5-carboxylic acid ≥97%

Proveedor: Apollo Scientific

OR510097-1G OR510097-250MG
APOSOR510097-1GEA 279 EUR
APOSOR510097-1G APOSOR510097-250MG
7-Nitro-1H-benzo[d][1,2,3]triazole-5-carboxylic acid ≥97%
7-Nitro-1H-benzo[d][1,2,3]triazole-5-carboxylic acid
Formula: C₇H₄N₄O₄
MW: 208,131 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 333767-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR