1-Amino-1H-pyrrole-2-carbonitrile hydrochloride 95%
Proveedor: Apollo Scientific
OR51360-1G
OR51360-5G
APOSOR51360-1GEA
116
EUR
APOSOR51360-1G
APOSOR51360-5G
1-Amino-1H-pyrrole-2-carbonitrile hydrochloride 95%
1-Amino-1H-pyrrole-2-carbonitrile hydrochloride
Formula:
C₅H₅N₃·ClH Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD13191739 Núm. CAS: 937046-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...