Methyl 2,4-dimethylbenzoate 95%

Proveedor: Apollo Scientific

OR52899-25G OR52899-5G
APOSOR52899-25GEA 48 EUR
APOSOR52899-25G APOSOR52899-5G
Methyl 2,4-dimethylbenzoate 95%
Methyl 2,4-dimethylbenzoate
Formula: C₁₀H₁₂O₂
MW: 164,201 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06203654
Núm. CAS: 23617-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR