5-(3,4-Dichlorophenyl)pentanoic acid 95%
Proveedor: Apollo Scientific
OR54560-100MG
OR54560-1G
OR54560-250MG
APOSOR54560-100MGEA
423
EUR
APOSOR54560-100MG
APOSOR54560-1G
APOSOR54560-250MG
5-(3,4-Dichlorophenyl)pentanoic acid 95%
5-(3,4-Dichlorophenyl)pentanoic acid
Formula:
C₁₁H₁₂Cl₂O₂ MW: 247,118 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09890961 Núm. CAS: 34658-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...