Methyl N-ethylglycinate hydrochloride 95%
Proveedor: Apollo Scientific
Sinónimos:
Methyl 2-(ethylamino)acetate hydrochloride, Methyl 2-(ethylamino)acetate, hydrochloride
OR70097-100MG
OR70097-1G
OR70097-250MG
APOSOR70097-100MGEA
116
EUR
APOSOR70097-100MG
APOSOR70097-1G
APOSOR70097-250MG
Methyl N-ethylglycinate hydrochloride 95%
Methyl N-ethylglycinate hydrochloride
Formula:
C₅H₁₁NO₂·ClH MW: 153,607 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16990638 Núm. CAS: 1121527-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...