(Oxan-4-ylmethyl)boronic acid 95%
Proveedor: Apollo Scientific
OR903431-1G
OR903431-250MG
APOSOR903431-1GEA
848
EUR
APOSOR903431-1G
APOSOR903431-250MG
(Oxan-4-ylmethyl)boronic acid 95%
(Oxan-4-ylmethyl)boronic acid
Formula:
C₆H₁₃BO₃ MW: 143,977 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD18861659 Núm. CAS: 1350513-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...