Methyl 3-tert-butylisoxazole-5-carboxylate 95%

Proveedor: Apollo Scientific

OR906938-100MG OR906938-1G OR906938-250MG
APOSOR906938-100MGEA 190 EUR
APOSOR906938-100MG APOSOR906938-1G APOSOR906938-250MG
Methyl 3-tert-butylisoxazole-5-carboxylate 95%
Methyl 3-tert-butylisoxazole-5-carboxylate
Formula: C₉H₁₃NO₃
MW: 183,204 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08752835
Núm. CAS: 133674-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR