5-Methyl-6-propoxypyridine-3-boronic acid 95%
Proveedor: Apollo Scientific
OR907376-1G
OR907376-5G
APOSOR907376-1GEA
219
EUR
APOSOR907376-1G
APOSOR907376-5G
5-Methyl-6-propoxypyridine-3-boronic acid 95%
5-Methyl-6-propoxypyridine-3-boronic acid
Formula:
C₉H₁₄BNO₃ MW: 195,023 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD18087691 Núm. CAS: 2096332-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...